
4-[5-(4,4,5,5-Tetramethyl[1,3,2]dioxaborolan-2-ylpiperazine-1-carboxylic acid tert-butyl ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:496786-98-2
:
Formula:C₂₀H₃₂BN₃O₄
MDL Number:MFCD04039875
:389.31 g/mol
Melting Pt:165…169 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together