Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Isoindoline-5-carboxylic acid hydrochloride
Isoindoline-5-carboxylic acid hydrochloride 95%
Isoindoline-5-carboxylic acid hydrochloride 95%
APOSOR960892-250MG
:
Apollo Scientific
:
Isoindoline-5-carboxylic acid hydrochloride 95%
APOSOR960892-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
199.64 g/mol
:
GHS07
:
Ambient
:
149353-72-0
:
C₉H₁₀ClNO₂
:
MFCD10699395