cis-1,2,3,6-Tetrahydrophthalic anhydride 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: cis-4-Cyclohexene-1,2-dicarboxylic anhydride
CAS Number:935-79-5
:
Formula:C₈H₈O₃
MDL Number:MFCD00005916
Molecular Weight:152.15 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together