Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo[1,2,4]triazolo[1,5-a]pyridine
5-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
5-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
APOSOR910006-1G
:
Apollo Scientific
:
5-Bromo[1,2,4]triazolo[1,5-a]pyridine 95%
APOSOR910006-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
198.0201 g/mol
:
Ambient
:
143329-58-2
:
C₆H₄BrN₃
:
MFCD10699201