

2-Cyclopenten-1-one 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:930-30-3
:
Formula:C₅H₆O
MDL Number:MFCD00001401
:82.1 g/mol
Boiling Pt:151…154 °C
Density:1,015 g/cm³ (25 °C)
Flash Pt:42 °C
Storage Temperature:Refrigerator
EINECS:213-213-0
UN:1224
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together