
1-n-Propyl-4-piperidone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23133-37-1
:
Formula:C₈H₁₅NO
MDL Number:MFCD00010433
:141.22 g/mol
Boiling Pt:56 °C/1mmHg
Density:0,936 g/cm³ (25 °C)
Flash Pt:76 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together