
5-(Piperidinomethyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5424-84-0
:
Formula:C₁₀H₁₅N₃OS
MDL Number:MFCD01689082
:225.31 g/mol
Melting Pt:224…225 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together