Order Entry
Ireland
ContactUsLinkComponent
2-Bromo-N,N-dimethylbenzenesulphonamide
Irritant
2-Bromo-N,N-dimethylbenzenesulphonamide
Catalog # APOSOR7770-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Bromo-N,N-dimethylbenzenesulphonamide
Catalog # APOSOR7770-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :264.139 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:65000-13-7
Formula:C₈H₁₀BrNO₂S
MDL Number:MFCD07368229