2,4,6-Trimethylphenylhydrazine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76195-82-9
:
Formula:C₉H₁₅ClN₂
MDL Number:MFCD00052268
:186.69 g/mol
Melting Pt:167…173 °C (dec.)
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Frequently Bought Together