Order Entry
Ireland
ContactUsLinkComponent
3'-Methoxy-[1,1'-biphenyl]-4-amine hydrochloride
Irritant
3'-Methoxy-[1,1'-biphenyl]-4-amine hydrochloride
Catalog # APOSOR7450-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3'-Methoxy-[1,1'-biphenyl]-4-amine hydrochloride
Catalog # APOSOR7450-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.71 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:207287-79-4
Formula:C₁₃H₁₃NO.HCl
MDL Number:MFCD07772428