
2-Bromophenyl methyl sulphone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33951-33-6
:
Formula:C₇H₇BrO₂S
MDL Number:MFCD07368924
:235.1 g/mol
Melting Pt:106…110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together