Order Entry
Ireland
ContactUsLinkComponent
[3-(Piperidin-1-ylmethyl)phenyl]amine 97%
Corrosive
Irritant
[3-(Piperidin-1-ylmethyl)phenyl]amine 97%
Catalog # APOSOR6791-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
[3-(Piperidin-1-ylmethyl)phenyl]amine 97%
Catalog # APOSOR6791-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.29 g/mol
Melting Pt:111…113 °C
Storage Temperature:Refrigerator
UN:2923
ADR:8+6.1,III
 :GHS05
 :GHS07
CAS Number:93138-55-7
Formula:C₁₂H₁₈N₂
MDL Number:MFCD04114502