

5-Chloro-2-methylbenzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7499-06-1
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00045842
:170.6 g/mol
Melting Pt:168…173 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together