2-((1-Benzylpiperidin-4-yl)methylene)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:120014-07-5
:
Formula:C₂₄H₂₇NO₃
MDL Number:MFCD08693144
:377.4761 g/mol
Storage Temperature:Ambient
Frequently Bought Together