Order Entry
Ireland
ContactUsLinkComponent
1,5-Diaminonaphthalene
Health
Environment
1,5-Diaminonaphthalene
Catalog # APOSOR60078-5G
Supplier:  Apollo Scientific
CAS Number:  
Health
Environment
1,5-Diaminonaphthalene
Catalog # APOSOR60078-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :158.2 g/mol
Boiling Pt:210 °C/5,25mmHg
Melting Pt:185…192 °C
Storage Temperature:Ambient
EINECS:218-817-8
UN:3077
ADR:9+EHS,III
 :GHS08
 :GHS09
CAS Number:2243-62-1
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004029