Order Entry
Ireland
ContactUsLinkComponent
4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid 98%
Irritant
4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid 98%
Catalog # APOSOR5711-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid 98%
Catalog # APOSOR5711-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:102…104 °C
 :367.21 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:850568-52-4
Formula:C₂₀H₂₂BNO₅
MDL Number:MFCD04973739