Order Entry
Ireland
ContactUsLinkComponent
1-Boc-6-methoxyindole-2-boronic acid 98%
Irritant
1-Boc-6-methoxyindole-2-boronic acid 98%
Catalog # APOSOR5551-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Boc-6-methoxyindole-2-boronic acid 98%
Catalog # APOSOR5551-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :291.11 g/mol
Melting Pt:100…103 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:850568-65-9
Formula:C₁₄H₁₈BNO₅
MDL Number:MFCD06659830