
1-Boc-5-cyanoindole-2-boronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:475102-15-9
:
Formula:C₁₄H₁₅BN₂O₄
MDL Number:MFCD04114578
:286.09 g/mol
Melting Pt:260…270 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together