
Methyl-4-Chloro-3-methylbenzoate
Catalog # APOSOR5430-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.62 g/mol
Boiling Pt:78 °C/0,06mmHg
Density:1,21 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:91367-05-4
Formula:C₉H₉ClO₂
MDL Number:MFCD00151838