

3-Chlorothiophenol 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2037-31-2
:
Formula:C₆H₅ClS
MDL Number:MFCD00004839
:144.62 g/mol
Boiling Pt:110 °C/30mmHg
Density:1,245 g/cm³ (25 °C)
Flash Pt:90 °C
Storage Temperature:Ambient
EINECS:218-010-0
UN:1760
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together