4-Methylcyclohexanone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:589-92-4
:
Formula:C₇H₁₂O
MDL Number:MFCD00001643
:112.17 g/mol
Boiling Pt:169…171 °C
Melting Pt:–41 °C
Density:0,914 g/cm³ (25 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
UN:2297
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together