
3,4-Dimethoxybenzylamine 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5763-61-1
:
Formula:C₉H₁₃NO₂
MDL Number:MFCD00008116
:167.21
Boiling Pt:281…284 °C
Density:1,109 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:227-287-7
UN:2735
ADR:8,III
:GHS05
Frequently Bought Together