
(S)-(-)-3-Acetamidopyrrolidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:114636-31-6
:
Formula:C₆H₁₂N₂O
MDL Number:MFCD00082748
:128.17 g/mol
Boiling Pt:70…72 °C/40mmHg
Melting Pt:52…57 °C
Storage Temperature:Refrigerator
UN:3259
ADR:8,III
:GHS05
Frequently Bought Together