
(S)-(+)-3-Amino-1-boc-piperidine
Catalog # APOSOR4588-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:200.28 g/mol
Density:1,02 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
CAS Number:625471-18-3
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD03094718