
4-Toluenethiol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:106-45-6
:
Formula:C₇H₈S
MDL Number:MFCD00004851
:124.21 g/mol
Boiling Pt:195 °C
Melting Pt:40…44 °C
Flash Pt:68 °C
Storage Temperature:Refrigerator
EINECS:203-399-1
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together