Order Entry
Ireland
ContactUsLinkComponent
2-(N-Methylamino)-3-hydroxymethylpyridine 98%
2-(N-Methylamino)-3-hydroxymethylpyridine 98%
Catalog # APOSOR42175-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(N-Methylamino)-3-hydroxymethylpyridine 98%
Catalog # APOSOR42175-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :138.1700 g/mol
Storage Temperature:Ambient
CAS Number:32399-12-5
Formula:C₇H₁₀N₂O
MDL Number:MFCD06254696