
1-n-Propyl-4-cyclopropylaminopiperidine 95+%
Catalog # APOSOR4114-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:182.31 g/mol
Boiling Pt:80 °C/2mmHg
:GHS07
Storage Temperature:Ambient
CAS Number:387358-45-4
Formula:C₁₁H₂₂N₂
MDL Number:MFCD01941332