Order Entry
Ireland
ContactUsLinkComponent
2-Methyl-4-hydroxymethylpyridine
Irritant
2-Methyl-4-hydroxymethylpyridine
Catalog # APOSOR40250-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methyl-4-hydroxymethylpyridine
Catalog # APOSOR40250-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :123.16 g/mol
Melting Pt:58…59 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:105250-16-6
Formula:C₇H₉NO
MDL Number:MFCD08062415