Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-7-methoxy-3-quinolinecarbaldehyde
2-Chloro-7-methoxy-3-quinolinecarbaldehyde
2-Chloro-7-methoxy-3-quinolinecarbaldehyde
2-Chloro-7-methoxy-3-quinolinecarbaldehyde
APOSOR40027-1G
:
Apollo Scientific
:
2-Chloro-7-methoxy-3-quinolinecarbaldehyde
APOSOR40027-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
221.64 g/mol
:
GHS07
:
Ambient
:
68236-20-4
:
C₁₁H₈ClNO₂
:
MFCD02227048