
indane-5-sulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52205-85-3
:
Formula:C₉H₉ClO₂S
MDL Number:MFCD05237217
:216.679 g/mol
Melting Pt:47,8…48.8 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together