Order Entry
Ireland
ContactUsLinkComponent
2,4,6-Triisopropylphenylboronic acid 98%
2,4,6-Triisopropylphenylboronic acid 98%
Catalog # APOSOR360494-25G
Supplier:  Apollo Scientific
CAS Number:  
2,4,6-Triisopropylphenylboronic acid 98%
Catalog # APOSOR360494-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :248.16 g/mol
Storage Temperature:Ambient
CAS Number:154549-38-9
Formula:C₁₅H₂₅BO₂
MDL Number:MFCD02683099