Order Entry
Ireland
ContactUsLinkComponent
1-Boc-indole-3-boronic acid
Irritant
1-Boc-indole-3-boronic acid
Catalog # APOSOR350387-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Boc-indole-3-boronic acid
Catalog # APOSOR350387-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :261.0814 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:181365-26-4
Formula:C₁₃H₁₆BNO₄
MDL Number:MFCD06656257