Order Entry
Ireland
ContactUsLinkComponent
1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid 98%
Irritant
1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid 98%
Catalog # APOSOR3422-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid 98%
Catalog # APOSOR3422-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :391.35 g/mol
Melting Pt:104…106 °C
 :GHS07
Storage Temperature:Freezer
CAS Number:335649-61-1
Formula:C₁₉H₃₀BNO₅Si
MDL Number:MFCD08689532