2-[(1-Methyl-3-oxo-3-phenyl-1-propenyl)amino]acetonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56464-51-8
:
Formula:C₁₂H₁₂N₂O
MDL Number:MFCD00102733
:200.24 g/mol
Storage Temperature:Ambient
Frequently Bought Together