Order Entry
Ireland
ContactUsLinkComponent
 
Methyl-2H-chromene-3-carboxylate 95%
Catalog # APOSOR32724-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-2H-chromene-3-carboxylate 95%
Catalog # APOSOR32724-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.2 g/mol
Storage Temperature:Ambient
CAS Number:36044-49-2
Formula:C₁₁H₁₀O₃
MDL Number:MFCD00202022