Order Entry
Ireland
ContactUsLinkComponent
Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate 95%
Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate 95%
Catalog # APOSOR32387-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate 95%
Catalog # APOSOR32387-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :264.67 g/mol
Storage Temperature:Ambient
CAS Number:129109-17-7
Formula:C₁₂H₉ClN₂O₃
MDL Number:MFCD00141141