1-(6-Chloro-3-pyridazinyl)piperidin-4-one 95%
Catalog # APOSOR32188-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:211.648 g/mol
Storage Temperature:Ambient
CAS Number:303149-95-3
Formula:C₉H₁₀ClN₃O
MDL Number:MFCD00665150