Order Entry
Ireland
ContactUsLinkComponent
Ethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate 95+%
Ethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate 95+%
Catalog # APOSOR318043-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate 95+%
Catalog # APOSOR318043-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.17 g/mol
Storage Temperature:Ambient
CAS Number:33458-27-4
Formula:C₇H₁₀N₂O₃
MDL Number:MFCD10688281