Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde
APOSOR30990-1G
:
Apollo Scientific
:
5-(3-Oxo-3-phenyl-1-propenyl)-2-furaldehyde
APOSOR30990-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
226.23 g/mol
:
Ambient
:
6575-75-3
:
C₁₄H₁₀O₃
:
MFCD01315261