N-(tert-Butoxycarbonyl)-L-pyroglutamic acid 95+%
Catalog # APOSOR309165-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:229.23 g/mol
Storage Temperature:Ambient
CAS Number:53100-44-0
Formula:C₁₀H₁₅NO₅
MDL Number:MFCD00672316