Order Entry
Ireland
ContactUsLinkComponent
 
Irritant
Triammonium citrate 97+%
Catalog # APOSOR30760-500G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Triammonium citrate 97+%
Catalog # APOSOR30760-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :243.22 g/mol
Melting Pt:185 °C
EINECS:222-394-5
 :GHS07
Storage Temperature:Ambient
CAS Number:3458-72-8
Formula:C₆H₁₇N₃O₇
MDL Number:MFCD00036406