(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:46022-05-3
:
Formula:C₈H₁₂O₄
MDL Number:MFCD00062973
:172.18 g/mol
Melting Pt:184 °C
Storage Temperature:Ambient
Frequently Bought Together