Order Entry
Ireland
ContactUsLinkComponent
N-Boc-cis-4-hydroxy-L-proline 95+%
Irritant
Environment
N-Boc-cis-4-hydroxy-L-proline 95+%
Catalog # APOSOR307072-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
N-Boc-cis-4-hydroxy-L-proline 95+%
Catalog # APOSOR307072-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :231.248 g/mol
Melting Pt:146 °C
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
 :GHS07
 :GHS09
CAS Number:87691-27-8
Formula:C₁₀H₁₇NO₅
MDL Number:MFCD02094406