
Benzyl-4-oxo-1,2,3,4-tetrahydropyridine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:185847-84-1
:
Formula:C₁₃H₁₃NO₃
MDL Number:MFCD08437477
:231.25 g/mol
Melting Pt:65…67 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together