1-(2,3-Dihydro-1H-inden-2-yl)-2-methylpropan-2-amine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1034457-07-2
:
Formula:C₁₃H₂₀ClN
MDL Number:MFCD22383947
:225.76 g/mol
Storage Temperature:Ambient
Frequently Bought Together