Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-[2-(Acetylamino)phenyl]acetamide
N-[2-(Acetylamino)phenyl]acetamide
N-[2-(Acetylamino)phenyl]acetamide
APOSOR30279-1G
:
Apollo Scientific
:
N-[2-(Acetylamino)phenyl]acetamide
APOSOR30279-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
Environmentally Preferable:
:
N,N'-(1,2-Phenylene)diacetamide
:
:
192.22 g/mol
:
Ambient
:
2050-85-3
:
C₁₀H₁₂N₂O₂
:
MFCD00092752