
5-Acetylindan
Catalog # APOSOR30257-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:85…87 °C/0,5mmHg
Density:1,06 g/cm³ (25 °C)
:160.22 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:4228-10-8
Formula:C₁₁H₁₂O
MDL Number:MFCD00021246