2,4,6-Trimethylaniline 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:88-05-1
:
Formula:C₉H₁₃N
MDL Number:MFCD00007740
:135.21 g/mol
Boiling Pt:233 °C
Melting Pt:–5 °C
Density:0,962 g/cm³ (25 °C)
Flash Pt:96 °C
Storage Temperature:Ambient
EINECS:201-794-3
UN:2810
ADR:6.1,II
:GHS06
:GHS08
Frequently Bought Together