
2-Chloro-6-methoxy-3-nitropyridine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:38533-61-8
:
Formula:C₆H₅ClN₂O₃
MDL Number:MFCD00130268
:188.57 g/mol
Melting Pt:78…80 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together