2,3-Pyrazinedicarboxylic anhydride 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2,3-Pyrazinecarboxylic anhydride
CAS Number:4744-50-7
:
Formula:C₆H₂N₂O₃
MDL Number:MFCD00179418
:150.09 g/mol
Melting Pt:210 °C (dec.)
EINECS:225-260-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together